Assay Details
Gene Family ID: | MIPF0000056, mir-148 |
Mature miRNA Sequence: | UCAGUGCAUGACAGAACUUGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-152-3p | MIMAT0000438 |
| Mouse | mmu-miR-152-3p | MIMAT0000162 |
| Rat | rno-miR-152-3p | MIMAT0000854 |
| Alligator mississippiensis | ami-miR-152-3p | MIMAT0038183 |
| Callithrix jacchus | cja-miR-152 | MIMAT0039463 |
| Chrysemys picta | cpi-miR-152-3p | MIMAT0037786 |
| View More Rows | ||
| Cricetulus griseus | cgr-miR-152-3p | MIMAT0023785 |
| Dog | cfa-miR-152 | MIMAT0006738 |
| Gorilla gorilla | ggo-miR-152 | MIMAT0024099 |
| Guinea pig | cpo-miR-152-3p | MIMAT0047033 |
| Pan troglodytes | ptr-miR-152 | MIMAT0008046 |
| Pig | ssc-miR-152 | MIMAT0013887 |
| Pongo pygmaeus | ppy-miR-152 | MIMAT0024356 |
| Pteropus alecto | pal-miR-152-3p | MIMAT0040107 |
| Rabbit | ocu-miR-152-3p | MIMAT0048268 |
| Rhesus monkey | mml-miR-152-3p | MIMAT0006214 |
| Sheep | oar-miR-152 | MIMAT0030035 |
| Tupaia chinensis | tch-miR-152 | MIMAT0036545 |
Stem-loop Details
Human
Mouse
Rat
Alligator mississippiensis
Callithrix jacchus
Chrysemys picta
Cricetulus griseus
Dog
Gorilla gorilla
Guinea pig
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Sheep
Tupaia chinensis

Genomic Map