Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 17 - 48036957 - 48036957 [+] on 48036957 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-10226 | MIMAT0041128 |
| Human | hsa-miR-152-3p | MIMAT0000438 |
| Human | hsa-miR-152-5p | MIMAT0026479 |
Stem-loop Details
Human

Genomic Map