Assay Details
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACCAUCGACCGUUGAGUGGAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-181c-3p | MIMAT0004559 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-181c-3p |
---|---|
Stem-loop Accession Number: | MI0000271 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AACCAUCGACCGUUGAGUGGAC |
Species: | Human |
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000007, mir-181 |
Mature miRNA Sequence: | AACCAUCGACCGUUGAGUGGAC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-181c-3p | MIMAT0004559 |