Assay Details
Amplicon Length: | 78 |
Chromosome Location: | Chr. 19 - 13874478 - 13874478 [+] on 13874478 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-181c-5p | MIMAT0000258 |
Human | hsa-miR-181c-3p | MIMAT0004559 |
Stem-loop Details
Human
