Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-28-5p | MIMAT0000085 |
| Mouse | mmu-miR-28a-5p | MIMAT0000653 |
| Rat | rno-miR-28-5p | MIMAT0000800 |
| Ateles geoffroyi | age-miR-28 | MIMAT0002351 |
| Bovine | bta-miR-28 | MIMAT0009272 |
| Dasypus novemcinctus | dno-miR-28-5p | MIMAT0047547 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-28 | MIMAT0002354 |
| Guinea pig | cpo-miR-28-5p | MIMAT0046894 |
| Horse | eca-miR-28-5p | MIMAT0013092 |
| Lagothrix lagotricha | lla-miR-28 | MIMAT0002358 |
| Macaca nemestrina | mne-miR-28 | MIMAT0002356 |
| Pan paniscus | ppa-miR-28 | MIMAT0002359 |
| Pan troglodytes | ptr-miR-28 | MIMAT0002353 |
| Pig | ssc-miR-28-5p | MIMAT0002136 |
| Pongo pygmaeus | ppy-miR-28 | MIMAT0002355 |
| Rabbit | ocu-miR-28-5p | MIMAT0048141 |
| Rhesus monkey | mml-miR-28-5p | MIMAT0002352 |
| Saguinus labiatus | sla-miR-28 | MIMAT0002357 |
| Tupaia chinensis | tch-miR-28-5p | MIMAT0036565 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-28 |
| Stem-loop Accession # | MI0000086 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000085
miRBase ID: hsa-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: Chr. 3 - 188688781 - 188688866 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-28a |
| Stem-loop Accession # | MI0000690 |
| Stem-loop Sequence | GGUCCCUACCUUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGAUUCUCCCACUAGAUUGUGAGCUGCUGGAGGGCAGGCACU |
| Chromosome Location | Chr. 16 - 24827855 - 24827940 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000653
miRBase ID: mmu-miR-28a-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: Chr. 16 - 24827855 - 24827940 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-28 |
| Stem-loop Accession # | MI0000861 |
| Stem-loop Sequence | GGUCCCUACCCGCAAGGAGCUCACAGUCUAUUGAGUUCCUUUUCUGAUUCUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000800
miRBase ID: rno-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: Chr. 11 - 79380523 - 79380608 [-] on Build Rnor_6.0 |
| Stem-loop ID | age-mir-28 |
| Stem-loop Accession # | MI0002648 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002351
miRBase ID: age-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | bta-mir-28 |
| Stem-loop Accession # | MI0009785 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGAGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009272
miRBase ID: bta-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | dno-mir-28 |
| Stem-loop Accession # | MI0038929 |
| Stem-loop Sequence | AAGGAGCUCACAGUCUAUUGAGUUUCCUUUUUGACUUUCCCACUAGAUUGUGAGCUCCUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047547
miRBase ID: dno-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | ggo-mir-28 |
| Stem-loop Accession # | MI0002651 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002354
miRBase ID: ggo-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | cpo-mir-28 |
| Stem-loop Accession # | MI0038585 |
| Stem-loop Sequence | AAGGAGCUCACAGUCUAUUGAGUUACUUUUAAGACUUUCCCACUAGAUUGGGAGCUCCUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046894
miRBase ID: cpo-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | eca-mir-28 |
| Stem-loop Accession # | MI0012840 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0013092
miRBase ID: eca-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | lla-mir-28 |
| Stem-loop Accession # | MI0002655 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002358
miRBase ID: lla-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | mne-mir-28 |
| Stem-loop Accession # | MI0002653 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUUCCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002356
miRBase ID: mne-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | ppa-mir-28 |
| Stem-loop Accession # | MI0002656 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002359
miRBase ID: ppa-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | ptr-mir-28 |
| Stem-loop Accession # | MI0002650 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002353
miRBase ID: ptr-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | ssc-mir-28 |
| Stem-loop Accession # | MI0002430 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUGUCUUCCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002136
miRBase ID: ssc-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | ppy-mir-28 |
| Stem-loop Accession # | MI0002652 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUACCUUUCUGAUUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002355
miRBase ID: ppy-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | ocu-mir-28 |
| Stem-loop Accession # | MI0039242 |
| Stem-loop Sequence | AAGGAGCUCACAGUCUAUUGAGUUGCCAUUCUGACUUCCCCACUAGAUUAUGAGCUCCUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048141
miRBase ID: ocu-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | mml-mir-28 |
| Stem-loop Accession # | MI0002649 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUUCCUUUGUGACUUUCCCACUAGAUUGUGAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002352
miRBase ID: mml-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | sla-mir-28 |
| Stem-loop Accession # | MI0002654 |
| Stem-loop Sequence | GGUCCUUGCCCUCAAGGAGCUCACAGUCUAUUGAGUUGCCUUUCUAACUUUCCCACUAGAUUGUAAGCUCCUGGAGGGCAGGCACU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002357
miRBase ID: sla-miR-28 Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
| Stem-loop ID | tch-mir-28 |
| Stem-loop Accession # | MI0031226 |
| Stem-loop Sequence | AAGGAGCUCACAGUCUAUUGAGUUGCCUUCCUGACUUUCUCACUAGAUUGUGAGCUCCUGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036565
miRBase ID: tch-miR-28-5p Mature miRNA Sequence: AAGGAGCUCACAGUCUAUUGAG Chromosome Location: NA |
More Information
Related Products

Genomic Map