Assay Details
Gene Family ID: | MIPF0000620, mir-1245 |
Mature miRNA Sequence: | AAGUGAUCUAAAGGCCUACAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1245a | MIMAT0005897 |
| Pan troglodytes | ptr-miR-1245 | MIMAT0007971 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map