Assay Details
Amplicon Length: | 77 |
Chromosome Location: | Chr. 2 - 188978495 - 188978495 [-] on 188978495 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1245a | MIMAT0005897 |
Human | hsa-miR-1245b-5p | MIMAT0019950 |
Human | hsa-miR-1245b-3p | MIMAT0019951 |
Stem-loop Details
Human
