Assay Details
Gene Family ID: | MIPF0000483, mir-1271 |
Mature miRNA Sequence: | CUUGGCACCUAGCAAGCACUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1271-5p | MIMAT0005796 |
| Gorilla gorilla | ggo-miR-1271 | MIMAT0024192 |
| Pan troglodytes | ptr-miR-1271 | MIMAT0007992 |
| Pongo pygmaeus | ppy-miR-1271 | MIMAT0016195 |
| Rhesus monkey | mml-miR-1271-5p | MIMAT0024315 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map