Assay Details
Gene Family ID: | MIPF0000591, mir-1278 |
Mature miRNA Sequence: | UAGUACUGUGCAUAUCAUCUAU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1278 | MIMAT0005936 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-1278 |
|---|---|
| Stem-loop Accession Number: | MI0006425 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGUACUGUGCAUAUCAUCUAU |
| Species: | Human |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000591, mir-1278 |
Mature miRNA Sequence: | UAGUACUGUGCAUAUCAUCUAU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1278 | MIMAT0005936 |