Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 1 - 193136301 - 193136301 [+] on 193136301 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1278 | MIMAT0005936 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 68 |
Chromosome Location: | Chr. 1 - 193136301 - 193136301 [+] on 193136301 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1278 | MIMAT0005936 |