Assay Details
Gene Family ID: | MIPF0000034, mir-130 |
Mature miRNA Sequence: | GCUCUGACGAGGUUGCACUACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-301b-5p | MIMAT0032026 |
| Guinea pig | cpo-miR-301b-5p | MIMAT0047144 |
| Pteropus alecto | pal-miR-301b-5p | MIMAT0040333 |
| Rabbit | ocu-miR-301b-5p | MIMAT0048366 |
Stem-loop Details
Human
Guinea pig
Pteropus alecto
Rabbit

Genomic Map