Assay Details
Gene Family ID: | MIPF0000229, mir-490 |
Mature miRNA Sequence: | CCAUGGAUCUCCAGGUGGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-490-5p | MIMAT0004764 |
| Mouse | mmu-miR-490-5p | MIMAT0017261 |
| Rat | rno-miR-490-5p | MIMAT0017356 |
| Anolis carolinensis | aca-miR-490-5p | MIMAT0021976 |
| Chicken | gga-miR-490-5p | MIMAT0026633 |
| Chrysemys picta | cpi-miR-490-5p | MIMAT0037899 |
| View More Rows | ||
| Dasypus novemcinctus | dno-miR-490-5p | MIMAT0047922 |
| Guinea pig | cpo-miR-490-5p | MIMAT0047283 |
| Horse | eca-miR-490-5p | MIMAT0012944 |
| Ophiophagus hannah | oha-miR-490-5p | MIMAT0036936 |
| Ornithorhynchus anatinus | oan-miR-490-5p | MIMAT0006810 |
| Pongo pygmaeus | ppy-miR-490-5p | MIMAT0015909 |
| Pteropus alecto | pal-miR-490-5p | MIMAT0040031 |
| Rabbit | ocu-miR-490-5p | MIMAT0048479 |
| Rhesus monkey | mml-miR-490-5p | MIMAT0006350 |
| Taeniopygia guttata | tgu-miR-490-5p | MIMAT0026996 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Chicken
Chrysemys picta
Dasypus novemcinctus
Guinea pig
Horse
Ophiophagus hannah
Ornithorhynchus anatinus
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Taeniopygia guttata

Genomic Map