Assay Details
Amplicon Length: | 96 |
Chromosome Location: | Chr. 7 - 136902971 - 136902971 [-] on 136902971 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-490-5p | MIMAT0004764 |
Human | hsa-miR-490-3p | MIMAT0002806 |
Stem-loop Details
Human
