Assay Details
Gene Family ID: | MIPF0000229, mir-490 |
Mature miRNA Sequence: | CCAUGGAUCUCCAGGUGGGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-490-5p | MIMAT0004764 |
Mouse | mmu-miR-490-5p | MIMAT0017261 |
Rat | rno-miR-490-5p | MIMAT0017356 |
Anolis carolinensis | aca-miR-490-5p | MIMAT0021976 |
Chicken | gga-miR-490-5p | MIMAT0026633 |
Chrysemys picta | cpi-miR-490-5p | MIMAT0037899 |
View More Rows | ||
Dasypus novemcinctus | dno-miR-490-5p | MIMAT0047922 |
Guinea pig | cpo-miR-490-5p | MIMAT0047283 |
Horse | eca-miR-490-5p | MIMAT0012944 |
Ophiophagus hannah | oha-miR-490-5p | MIMAT0036936 |
Ornithorhynchus anatinus | oan-miR-490-5p | MIMAT0006810 |
Pongo pygmaeus | ppy-miR-490-5p | MIMAT0015909 |
Pteropus alecto | pal-miR-490-5p | MIMAT0040031 |
Rabbit | ocu-miR-490-5p | MIMAT0048479 |
Rhesus monkey | mml-miR-490-5p | MIMAT0006350 |
Taeniopygia guttata | tgu-miR-490-5p | MIMAT0026996 |
Stem-loop Details
Human
Mouse
Rat
Anolis carolinensis
Chicken
Chrysemys picta
Dasypus novemcinctus
Guinea pig
Horse
Ophiophagus hannah
Ornithorhynchus anatinus
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Taeniopygia guttata

More Information
Related Products
miRBase Alias:
Rat: rno-miR-490* (v18)
Ochrobactrum anthropi: oan-miR-490 (v18)
Ochrobactrum anthropi: oan-miR-490 (v18)
