Assay Details
Amplicon Length: | 90 |
Chromosome Location: | Chr. 6 - 36422043 - 36422043 [+] on 36422043 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-490-5p | MIMAT0017261 |
| Mouse | mmu-miR-490-3p | MIMAT0003780 |
Stem-loop Details
Mouse

Genomic Map