Assay Details
Amplicon Length: | 75 |
Chromosome Location: | Chr. 4 - 63342797 - 63342797 [-] on 63342797 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-490-3p | MIMAT0012823 |
| Rat | rno-miR-490-5p | MIMAT0017356 |
Stem-loop Details
Rat

Genomic Map