Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | GAAAGCGCUUCUCUUUAGAGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-518f-3p | MIMAT0002842 |
| Gorilla gorilla | ggo-miR-518f-3p | MIMAT0036417 |
| Pan troglodytes | ptr-miR-518f | MIMAT0008191 |
| Pongo pygmaeus | ppy-miR-518f | MIMAT0015958 |
| Pongo pygmaeus | ppy-miR-518f | MIMAT0015957 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus

Genomic Map