Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. 19 - 53699775 - 53699775 [-] on 53699775 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-518f-3p | MIMAT0002842 |
Human | hsa-miR-518f-5p | MIMAT0002841 |
Stem-loop Details
Human
