Assay Details
Gene Family ID: | MIPF0000623, mir-561 |
Mature miRNA Sequence: | AUCAAGGAUCUUAAACUUUGCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-561-5p | MIMAT0022706 |
| Gorilla gorilla | ggo-miR-561 | MIMAT0049113 |
| Nomascus leucogenys | nle-miR-561 | MIMAT0049464 |
Stem-loop Details
Human
Gorilla gorilla
Nomascus leucogenys

Genomic Map