Assay Details
Amplicon Length: | 75 |
Chromosome Location: | Chr. 2 - 188297288 - 188297288 [+] on 188297288 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-561-5p | MIMAT0022706 |
Human | hsa-miR-561-3p | MIMAT0003225 |
Stem-loop Details
Human
