Assay Details
Gene Family ID: | MIPF0000649, mir-1296 |
Mature miRNA Sequence: | GAGUGGGGCUUCGACCCUAACC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1296-3p | MIMAT0026637 |
| Rhesus monkey | mml-miR-1296-3p | MIMAT0027114 |
Stem-loop Details
Human
Rhesus monkey

Genomic Map