Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 10 - 63373258 - 63373258 [-] on 63373258 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1296-5p | MIMAT0005794 |
| Human | hsa-miR-1296-3p | MIMAT0026637 |
Stem-loop Details
Human

Genomic Map