Assay Details
Mature miRNA Sequence: | GUAUACACCUGAUAUGUGUAUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4789-5p | MIMAT0019959 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-4789-5p |
|---|---|
| Stem-loop Accession Number: | MI0017436 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUAUACACCUGAUAUGUGUAUG |
| Species: | Human |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Mature miRNA Sequence: | GUAUACACCUGAUAUGUGUAUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4789-5p | MIMAT0019959 |