Assay Details
Amplicon Length: | 138 |
Chromosome Location: | Chr. 3 - 175369247 - 175369247 [-] on 175369247 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4789-3p | MIMAT0019960 |
| Human | hsa-miR-4789-5p | MIMAT0019959 |
Stem-loop Details
Human

Genomic Map