Assay Details
Gene Family ID: | MIPF0000039, mir-34 |
Mature miRNA Sequence: | UAGGCAGUGUCAUUAGCUGAUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-34b-5p | MIMAT0000685 |
Stem-loop Details
Human
More Information
Related Products
miRBase Alias:
Human: hsa-miR-34b (v9.2)
Human: hsa-miR-34b* (v17)
Human: hsa-miR-34b* (v17)

Genomic Map