Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 11 - 111512543 - 111512543 [-] on 111512543 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-34b-3p | MIMAT0004676 |
| Human | hsa-miR-34b-5p | MIMAT0000685 |
Stem-loop Details
Human

Genomic Map