Assay Details
Gene Family ID: | MIPF0000512, mir-562 |
Mature miRNA Sequence: | AAAGUAGCUGUACCAUUUGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-562 | MIMAT0003226 |
Pan troglodytes | ptr-miR-562 | MIMAT0008242 |
Pongo pygmaeus | ppy-miR-562 | MIMAT0016007 |
Rhesus monkey | mml-miR-562 | MIMAT0006428 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey
