Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 2 - 232172351 - 232172351 [+] on 232172351 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-562 | MIMAT0003226 |
Stem-loop Details
Human
