Assay Details
| Gene Family ID: | MIPF0000317, mir-548 | 
| Mature miRNA Sequence: | CAAAAACCACAGUUUCUUUUGC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-548d-3p | MIMAT0003323 | 
Stem-loop Details
Human
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-548d-3p | 
|---|---|
| miRBase Accession Number: | MI0003668 | 
| miRBase Version: | v22.1 | 
| Mature miRNA Sequence: | CAAAAACCACAGUUUCUUUUGC | 
| Species: | Human | 
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor | 
| Gene Family ID: | MIPF0000317, mir-548 | 
| Mature miRNA Sequence: | CAAAAACCACAGUUUCUUUUGC | 
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-548d-3p | MIMAT0003323 | 
 Back To Top
 Back To Top