Assay Details
Amplicon Length: | 91 |
Chromosome Location: | Chr. 17 - 67471984 - 67471984 [-] on 67471984 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548aa | MIMAT0018447 |
| Human | hsa-miR-548d-5p | MIMAT0004812 |
| Human | hsa-miR-548d-3p | MIMAT0003323 |
Stem-loop Details
Human

Genomic Map