Assay Details
Gene Family ID: | MIPF0000109, mir-186 |
Mature miRNA Sequence: | CAAAGAAUUCUCCUUUUGGGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-186-5p | MIMAT0000456 |
| Mouse | mmu-miR-186-5p | MIMAT0000215 |
| Rat | rno-miR-186-5p | MIMAT0000863 |
| Bovine | bta-miR-186 | MIMAT0003818 |
| Callithrix jacchus | cja-miR-186 | MIMAT0039388 |
| Daubentonia madagascariensis | dma-miR-186 | MIMAT0049271 |
| View More Rows | ||
| Dog | cfa-miR-186 | MIMAT0006694 |
| Goat | chi-miR-186-5p | MIMAT0036010 |
| Horse | eca-miR-186 | MIMAT0012956 |
| Nomascus leucogenys | nle-miR-186 | MIMAT0049448 |
| Ornithorhynchus anatinus | oan-miR-186-5p | MIMAT0006931 |
| Papio hamadryas | pha-miR-186 | MIMAT0049515 |
| Pongo pygmaeus | ppy-miR-186 | MIMAT0015782 |
| Rhesus monkey | mml-miR-186-5p | MIMAT0006220 |
| Tupaia chinensis | tch-miR-186-5p | MIMAT0036622 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Daubentonia madagascariensis
Dog
Goat
Horse
Nomascus leucogenys
Ornithorhynchus anatinus
Papio hamadryas
Pongo pygmaeus
Rhesus monkey
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-186 (v17)
Mouse: mmu-miR-186 (v17)
Rat: rno-miR-186 (v18)
Macaca mulatta: mml-miR-186 (v19)
Ochrobactrum anthropi: oan-miR-186 (v18)
Mouse: mmu-miR-186 (v17)
Rat: rno-miR-186 (v18)
Macaca mulatta: mml-miR-186 (v19)
Ochrobactrum anthropi: oan-miR-186 (v18)

Genomic Map