Assay Details
Amplicon Length: | 68 |
Chromosome Location: | Chr. 2 - 263873510 - 263873510 [-] on 263873510 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-186-5p | MIMAT0000863 |
| Rat | rno-miR-186-3p | MIMAT0017143 |
Stem-loop Details
Rat

Genomic Map