Assay Details
Gene Family ID: | MIPF0000690, mir-943 |
Mature miRNA Sequence: | CUGACUGUUGCCGUCCUCCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-943 | MIMAT0004986 |
| Pan troglodytes | ptr-miR-943 | MIMAT0008349 |
Stem-loop Details
Human
Pan troglodytes

Genomic Map