Assay Details
Gene Family ID: | MIPF0000690, mir-943 |
Mature miRNA Sequence: | CUGACUGUUGCCGUCCUCCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-943 | MIMAT0004986 |
Pan troglodytes | ptr-miR-943 | MIMAT0008349 |
Stem-loop Details
Human
Pan troglodytes
