Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UCAUUCACGGACAACACUUUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-382-3p | MIMAT0004691 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-382-3p |
|---|---|
| miRBase Accession Number: | MI0000799 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCAUUCACGGACAACACUUUUU |
| Species: | Mouse |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UCAUUCACGGACAACACUUUUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-382-3p | MIMAT0004691 |