Assay Details
Amplicon Length: | 61 |
Chromosome Location: | Chr. 12 - 109733976 - 109733976 [-] on 109733976 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-134-3p | MIMAT0016985 |
| Mouse | mmu-miR-134-5p | MIMAT0000146 |
| Mouse | mmu-miR-382-5p | MIMAT0000747 |
| Mouse | mmu-miR-382-3p | MIMAT0004691 |
Stem-loop Details
Mouse

Genomic Map