Assay Details
Gene Family ID: | MIPF0000604, mir-1284 |
Mature miRNA Sequence: | UCUAUACAGACCCUGGCUUUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1284 | MIMAT0005941 |
Pan troglodytes | ptr-miR-1284 | MIMAT0008003 |
Pongo pygmaeus | ppy-miR-1284 | MIMAT0016206 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
