Assay Details
Gene Family ID: | MIPF0000604, mir-1284 |
Mature miRNA Sequence: | UCUAUACAGACCCUGGCUUUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1284 | MIMAT0005941 |
| Pan troglodytes | ptr-miR-1284 | MIMAT0008003 |
| Pongo pygmaeus | ppy-miR-1284 | MIMAT0016206 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map