Assay Details
Mature miRNA Sequence: | CAAAUCCUUGCCCGGGUGCAUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3560 | MIMAT0017829 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-3560 |
---|---|
miRBase Accession Number: | MI0015422 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAAAUCCUUGCCCGGGUGCAUU |
Species: | Rat |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | CAAAUCCUUGCCCGGGUGCAUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3560 | MIMAT0017829 |