Assay Details
Amplicon Length: | 94 |
Chromosome Location: | Chr. X - 16118123 - 16118123 [+] on 16118123 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3560 | MIMAT0017829 |
Rat | rno-miR-501-5p | MIMAT0003116 |
Rat | rno-miR-501-3p | MIMAT0017198 |
Stem-loop Details
Rat
