Assay Details
Gene Family ID: | MIPF0001188, mir-1188 |
Mature miRNA Sequence: | CGAGGCUCCCCACCACA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-1188-3p | MIMAT0017855 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-1188-3p |
---|---|
miRBase Accession Number: | MI0015443 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CGAGGCUCCCCACCACA |
Species: | Rat |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0001188, mir-1188 |
Mature miRNA Sequence: | CGAGGCUCCCCACCACA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-1188-3p | MIMAT0017855 |