Assay Details
Amplicon Length: | 116 |
Chromosome Location: | Chr. 6 - 133733823 - 133733823 [+] on 133733823 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-1188-5p | MIMAT0017854 |
Rat | rno-miR-1188-3p | MIMAT0017855 |
Stem-loop Details
Rat
