Assay Details
Gene Family ID: | MIPF0001166, mir-3578 |
Mature miRNA Sequence: | AAGUGUUGUCCGUGAAUGAUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3592 | MIMAT0017895 |
Stem-loop Details
Rat

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | rno-miR-3592 |
---|---|
miRBase Accession Number: | MI0015473 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | AAGUGUUGUCCGUGAAUGAUU |
Species: | Rat |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0001166, mir-3578 |
Mature miRNA Sequence: | AAGUGUUGUCCGUGAAUGAUU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-3592 | MIMAT0017895 |