Assay Details
Amplicon Length: | 97 |
Chromosome Location: | Chr. 6 - 133883927 - 133883927 [+] on 133883927 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3592 | MIMAT0017895 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 97 |
Chromosome Location: | Chr. 6 - 133883927 - 133883927 [+] on 133883927 Build Rnor_6.0 |
Species: | Rat |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-3592 | MIMAT0017895 |