Assay Details
Gene Family ID: | MIPF0000026, mir-218 |
Mature miRNA Sequence: | AAACAUGGUUCCGUCAAGCAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-218a-1-3p | MIMAT0017162 |
Stem-loop Details
Rat
More Information
Related Products
miRBase Alias:
Rat: rno-miR-218-1* (v16)
Rat: rno-miR-218a-1* (v18)
Rat: rno-miR-218a-1* (v18)

Genomic Map