Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. 14 - 66922739 - 66922739 [+] on 66922739 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-218a-1-3p | MIMAT0017162 |
| Rat | rno-miR-218a-5p | MIMAT0000888 |
| Rat | rno-miR-218b | MIMAT0017838 |
Stem-loop Details
Rat

Genomic Map