Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAACCGCAGUAACUUUUGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548at-3p | MIMAT0022278 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-548at-3p |
---|---|
miRBase Accession Number: | MI0019137 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | CAAAACCGCAGUAACUUUUGU |
Species: | Human |
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | CAAAACCGCAGUAACUUUUGU |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548at-3p | MIMAT0022278 |