Assay Details
Amplicon Length: | 179 |
Chromosome Location: | Chr. 17 - 42494491 - 42494491 [-] on 42494491 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548at-5p | MIMAT0022277 |
Human | hsa-miR-548at-3p | MIMAT0022278 |
Stem-loop Details
Human
