Assay Details
Amplicon Length: | 73 |
Chromosome Location: | Chr. 3 - 120395461 - 120395461 [+] on 120395461 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-198 | MIMAT0000228 |
Stem-loop Details
Human
