Assay Details
Gene Family ID: | MIPF0000090, mir-198 |
Mature miRNA Sequence: | GGUCCAGAGGGGAGAUAGGUUC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-198 | MIMAT0000228 |
Stem-loop Details
Human

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-198 |
---|---|
miRBase Accession Number: | MI0000240 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GGUCCAGAGGGGAGAUAGGUUC |
Species: | Human |
Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000090, mir-198 |
Mature miRNA Sequence: | GGUCCAGAGGGGAGAUAGGUUC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-198 | MIMAT0000228 |